Mouse CCL7 - MCP3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0393
76236-876EA 760.6 USD
76236-876
Mouse CCL7 - MCP3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CCL7 - MCP3 AccuSignal ELISA Kit
  • Natural and recombinant mouse CCL7.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: Q24-P97
Chemokine(C-C motif) ligand 7(CCL7) is a small cytokine known as a chemokine that was previously called monocyte-specific chemokine 3(MCP3). It belongs to the C-C chemokine family. By fluorescence in situ hybridization, mapped the MCP3 gene to chromosome 17q11.2-q12. MCP3 was identified as a physiologic substrate of gelatinase A. Cleaved MCP3 binds to CC-chemokine receptors-1, -2, and -3, but no longer induces calcium fluxes or promotes chemotaxis, and instead acts as a general chemokine antagonist that dampens inflammation, suggested that matrix metalloproteinases are both effectors and regulators of the inflammatory response.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse CCL7 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CCL7 standard solution and each sample be measured in duplicate.