Human PDGF-AB ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0289
76236-774EA 760.6 USD
76236-774
Human PDGF-AB ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human PDGF-AB AccuSignal ELISA Kit
  • Natural and recombinant human PDGF-AB.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: (S87-T211)+(S82-T190)
The platelet-derived growth factor(PDGF) is a mitogen derived from human platelets consisting of two related polypeptides termed A and B chains. The genes for PDGF A chain, B chain/c-sis, and the PDGF receptor are expressed in human malignant glioma cell lines. Normal human endothelial cells in culture express the B chain of PDGF, and that endothelial-derived PDGF B chain is synthesized as a predicted precursor polypeptide of Mr 27,281. The entire B chain of PDGF is highly(96%) homologous to a portion of p28sis, the transforming protein of simian sarcoma virus(SSV). It has been suggested that p28sis exerts its transforming potential by mimicking the growth promoting activity of PDGF and stimulating the cell in an autocrine manner. PDGF A-chain precursor polypeptide is assigned to the proximal long arm of chromosome 7, band q11.23. The human homolog(PDGF B-chain/c-sis) of the transforming gene of simian sarcoma virus is assigned to chromosome 22. The standard product used in this kit is recombinant human PDGF-AB with the molecular mass of 27KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human PDGF-AB standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human PDGF-AB standard solution and each sample be measured in duplicate.