Mouse OPG ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0286
76236-768EA 760.6 USD
76236-768
Mouse OPG ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse OPG AccuSignal ELISA Kit
  • Natural and recombinant mouse OPG.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E22-L401
Osteoprotegerin(OPG) is identical to osteoclastogenesis inhibitory factor(OCIF), a soluble member of the tumor-necrosis factor receptor family that inhibits osteoclastogenesis. OPG is considered to play an important role in the regulation of bone resorption by modifying osteoclast differentiation. Osteoprotegerin is an independent risk factor for the progression of atherosclerosis and onset of cardiovascular disease. It can act as a soluble factor in the regulation of bone mass and imply a utility for OPG in the treatment of osteoporosis associated with increased osteoclast activity. OPG system may play a critical role in the development of osteolytic bone disease in multiple myeloma and that targeting this system may have therapeutic potential. OPG also plays a role in the vascular system.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 6000pg/ml, 3000pg/ml, 1500pg/ml, 750pg/ml, 419pg/ml, 187.5pg/ml, 93.7pg/m mouse OPG standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(EDTA) to each empty well. It is recommended that each mouse OPG standard solution and each sample be measured in duplicate.