Human IL-11 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0232
76236-688EA 760.6 USD
76236-688
Human IL-11 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-11 AccuSignal ELISA Kit
  • Natural and recombinant human IL-11.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: P22-L199
Interleukin 11(IL-11) is a protein that in humans is encoded by the IL11 gene. The protein encoded by this gene is a member of the gp130 family of cytokines. This gene is mapped to 19q13.42. It is a key regulator of multiple events in hematopoiesis, most notably the stimulation of megakaryocyte maturation. IL-11 has been demonstrated to improve platelet recovery after chemotherapy-induced thrombocytopenia, induce acute phase proteins, modulate antigen-antibody responses, participate in the regulation of bone cell proliferation and differentiation and could be use as a therapeutic for osteoporosis. What's more, IL-11 can stimulate the growth of certain lymphocytes and it has functions in many other tissues, including the brain, gut, testis and bone. In transgenic mice, overexpression of the human IL11 gene resulted in the stimulation of bone formation to increase cortical thickness and strength of long bones.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human IL-11 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernatants, serum or plasma(heparin, EDTA, citrate) to each empty well. We recommend that each human IL-11 standard solution and each sample is measured in duplicate.