Mouse Activin A ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0142
76236-588EA 679.1 USD
76236-588
Mouse Activin A ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Activin A AccuSignal ELISA Kit
  • Natural and recombinant mouse Activin A.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: G311-S427
Activin A is a homodimer of 14kDa beta-A. Activin A, a cytokine member of the transforming growth factor-beta superfamily, is expressed locally by the mesenchymal component of the hemopoietic microenvironment. Its expression is regulated on the mRNA level by different cytokines, and the biological activity of the protein is tightly controlled by several inhibitory molecules. Inhibins and activins are members of the transforming growth factor beta superfamily and are known to modulate the growth and differentiation of several cell types. Inhibins and activins inhibit and activate, respectively, the secretion of follitropin by the pituitary gland. Inhibins/activins are involved in regulating a number of diverse functions such as hypothalamic and pituitary hormone secretion, gonadal hormone secretion, germ cell development and maturation, erythroid differentiation, insulin secretion, nerve cell survival, embryonic axial development or bone growth, depending on their subunit composition. Inhibins appear to oppose the functions of activins. The standard product used in this kit is recombinant Activin A, which is composed of two single chains of 116 amino acids with the molecular mass of 26KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse Activin A standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum, plasma(heparin, EDTA) or saliva to each empty well. It is recommended that each mouse Activin A standard solution and each sample be measured in duplicate.