FACS and IF Staining Buffer with FBS

Supplier: Rockland Immunochemical

MB-086-0500
76236-550EA 106.63 USD
76236-550
FACS and IF Staining Buffer with FBS
Buffers Microscopy Buffers

Fluorescent Activated Cell Sorting (FACS) and Immunofluorescence (IF) staining buffer with FBS


  • Cell staining, washing and dilution
  • Flow cytometry and immunofluorescence
  • Contains 2% calf bovine serum as carrier and stabilizer


Fluorescent Activated Cell Sorting (FACS) and Immunofluorescence (IF) staining buffer is a diluent buffer optimized for use in flow cytometry and immunofluorescence assay of viable and/or fixed cells. FACS and IF staining, washing buffer contains 2% calf bovine serum as carrier and stabilizer to reduce non-specific binding of antibodies and fluorochrome reagents to targets cells. Metabolic inhibitors and NaN3 are also included in the flow cytometry and immunofluorescence buffer to prevent patching, capping of surface antigen and bacterial growth.


Application Note: Fluorescent Activated Cell Sorting (FACS) and Immunofluorescence (IF) staining buffer is adequate for cell staining, washing and dilution in flow cytometry and immunofluorescence experiments.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR