Tech-Med® Wartenberg Pinwheels, Dukal
Supplier: Dukal
7020
76236-498EA 18.02 USD
76236-498
Tech-Med® Wartenberg Pinwheels, Dukal
Clinical Diagnostic Supplies Clinical Instruments
Also known as a Wartenberg Neurowheel, the instrument is designed to test nerve reactions (sensitivity) as it rolled systematically across the skin.
  • Chrome finish
  • 7.5" overall height