Express Transfer Buffer
Supplier: Rockland Immunochemical
MB-024-1000
76236-400EA 99.25 USD
76236-400
Express Transfer Buffer
Buffers Electrophoresis Buffers Transfer Buffers
Express Transfer Buffer
  • Western blot transfer solution
  • Alternative to tris-glycine
  • Lower background
Western Blotting is the method used for immunodetection for the detection of proteins. Blotting is a method for transferring biological molecules (e.g., proteins, nucleic acid fragments) from a gel matrix to a membrane support for the subsequent detection of those molecules.
Application Note: Express Transfer Buffer is a alternative to tris-glycine as a transfer buffer in the western blot. It may run at an elevated temperature but gives markedly lower background.