QuantiFluo™ Diamine Oxidase Assay Kit, BioAssay Systems
Supplier: BIOASSAY SYSTEMS
QFDO-100
76236-146EA 709.21 USD
76236-146
QuantiFluo™ Diamine Oxidase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of diamine oxidase in biological samples (example: serum, plasma and more).
  • Fast and sensitive, convenient
  • Use of 10 μl sample
  • Linear detection range 0.5 to 6 U/l for 30 min reaction at 25 °C
  • High-throughput
  • "Add-mix-read" type assay
  • Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day
  • Method: FL530/585 nm, samples: Serum, Plasma
The procedure involves adding a single working reagent, and reading the fluorescence at 0 and 30 minutes. Room temperature assay, no 37 °C heater is needed. Diamine oxidase (DAO) also known as histaminase or amine oxidase (copper containing), is an enzyme involved in the metabolism, oxidation, and inactivation of histamine in animals. Highest content is observed in the digestive tract and placenta. An imbalance between histamine intake and the capacity for histamine degradation can lead to histamine intolerance (HIT). Measuring DAO activity in serum can be useful in diagnosing HIT.
Non-radioactive, fluorimetric DAO assay is based on the oxidation of putrescine to pyrroline plus NH3 and H₂O₂. The generated H₂O₂ is then used by HRP to oxidize a dye making it fluorescent. The increase in fluorescence at λex/em = 530/585 nm is directly proportional to the enzyme activity.