Mouse LBP ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0750
76235-964EA 688.51 USD
76235-964
Mouse LBP ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse LBP AccuSignal ELISA Kit
  • Natural and recombinant mouse LBP.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: G25-V481
Lipopolysaccharide binding protein is a protein that in humans is encoded by the LBP gene. This gene is mapped to 20q11.23. LBP is a soluble acute-phase protein that binds to bacterial lipopolysaccharide(or LPS) to elicit immune responses by presenting the LPS to important cell surface pattern recognition receptors called CD14 and TLR4. It is present in the cerebrospinal fluid of patients with pneumococcal meningitis. The protein encoded by this gene is involved in the acute-phase immunologic response to gram-negative bacterial infections. LBP is made in the liver during the acute phase of infections and is thought to function as a carrier for LPS and to help control LPS-dependent monocyte responses.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 100ng/ml, 50ng/ml, 25ng/ml, 1.25ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml mouse LBP standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse LBP standard solution and each sample is measured in duplicate.