Human FOLR1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0731
76235-942EA 760.6 USD
76235-942
Human FOLR1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human FOLR1 AccuSignal ELISA Kit
  • Natural and recombinant human FOLR1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: R25-S234
Folate receptor 1 adult is a protein that in humans is encoded by the FOLR1 gene. The protein encoded by this gene is a member of the folate receptor(FOLR) family. It is mapped to 11q13.4. FOLR1 expression in Jurkat cells facilitated MBG or EBO entry, and FR-blocking reagents inhibited infection of MBG or EBO. This gene product is a secreted protein that either anchors to membranes via a glycosyl-phosphatidylinositol linkage or exists in a soluble form. The FOLR1 gene encodes the adult folate receptor, or folate-binding protein(FBP), which has a high affinity for folic acid and for several reduced folic acid derivatives, and mediates delivery of 5-methyltetrahydrofolate to the interior of cells. FOLR1 is also an important regulator of milk protein synthesis.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 3000pg/ml, 1500pg/ml, 750pg/ml, 419pg/ml, 188pg/ml, 94pg/ml, 46.9pg/ml human FOLR1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA), saliva, urine or human milk to each empty well. We recommend that each human FOLR1 standard solution and each sample is measured in duplicate.