Mouse HGF ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0710
76235-900EA 760.6 USD
76235-900
Mouse HGF ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse HGF AccuSignal ELISA Kit
  • Natural and recombinant mouse HGF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q33-R495 (α)&V496-L728 (β)
Hepatocyte growth factor(HGF) is the most potent mitogen for mature parenchymal hepatocytes in primary culture, and seems to be a hepatotrophic factor that acts as a trigger for liver regeneration after partial hepatectomy and liver injury. HGF has a relative molecular mass(Mr) of 82,000 and is a heterodimer composed of a large alpha-subunit of Mr 69,000 and a small beta-subunit of Mr 34,000. The protein consists of 728 amino acid residues, including a possible signal peptide at the N-terminus. HGF may serve as a paracrine mediator to control placental development and growth. This growth factor may play an important role as a paracrine mediator of the proliferation of melanocytes and endothelial cells, as well as cells of epithelial origin. The gene encoding the human HGF is assigned to human chromosome 7.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse HGF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(EDTA, citrate) to each empty well. We recommend that each mouse HGF standard solution and each sample is measured in duplicate.