Human CD13 - Aminopeptidase N ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0709
76235-898EA 760.6 USD
76235-898
Human CD13 - Aminopeptidase N ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CD13 - Aminopeptidase N AccuSignal ELISA Kit
  • Natural and recombinant human CD13.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: K69-K967
Alanine aminopeptidase, also known as ANPEP or CD13. is an enzyme that is used as a biomarker to detect damage to the kidneys, and that may be used to help diagnose certain kidney disorders. It is mapped to 15q26.1. Aminopeptidase N is located in the small-intestinal and renal microvillar membrane, and also in other plasma membranes. In the small intestine, Aminopeptidase N plays a role in the final digestion of peptides generated from hydrolysis of proteins by gastric and pancreatic proteases, and it is also thought to be involved in the metabolism of regulatory peptides by diverse cell types, including small intestinal and renal tubular epithelial cells, macrophages, granulocytes, and synaptic membranes from the CNS. Petrovic et al showed that CD13 was required for endothelial cell invasion in response to bradykinin. Inhibition of CD13 abrogated internalization of bradykinin receptor B2 and reduced endothelial cell motility.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20,000pg/ml, 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human CD13 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CD13 standard solution and each sample be measured in duplicate.