Mouse IL-17RC ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0681
76235-842EA 760.6 USD
76235-842
Mouse IL-17RC ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Interleukin-17RC AccuSignal ELISA Kit
  • Natural and recombinant mouse IL-17RC.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E22-R440
Interleukin-17 receptor C is a protein that in humans is encoded by the IL17RC gene. By genomic sequence analysis, IL17RC gene was mapped to chromosome 3p25.3-p24.1. This gene encodes a single-pass transmembrane protein that shares limited similarity with the interleukin-17 receptor. Multiple alternatively spliced transcript variants encoding different isoforms have been detected for this gene, but the full-length nature of only three have been determined to date. The biologic activity of IL-17 is dependent on a complex composed of IL-17RA and IL-17RC, suggesting a new paradigm for understanding the interactions between the expanded family of IL-17 ligands and their receptors.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse IL-17RC standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse IL-17RC standard solution and each sample be measured in duplicate.