Rat EGF ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0584
76235-708EA 760.6 USD
76235-708
Rat EGF ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat EGF AccuSignal ELISA Kit
  • Natural and recombinant rat EGF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: N974-R1026
Epidermal growth factor(EGF) is a growth factor that plays an important role in the regulation of cell growth, proliferation and differentiation by binding to its receptor EGFR. EGF locus is mapped to 4q21-4qter. Human EGF is a 6045-Da protein with 53 amino acid residues and three intramolecular disulfide bones. EGF results in cellular proliferation, differentiation, and survival while plays a physiological role in the maintenance of oro-esophageal and gastric tissue integrity.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml rat EGF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum, plasma(heparin, EDTA), tissue homogenates or urine to each empty well. It is recommended that each rat EGF standard solution and each sample be measured in duplicate.