Human Endothelin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0579
76235-698EA 760.6 USD
76235-698
Human Endothelin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Endothelin AccuSignal ELISA Kit
  • Natural and recombinant human Endothelin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: human peptide; Immunogen sequence: C53-W73
The endothelins is a family of structurally and pharmacologically distinct peptides, which has been identified and sequenced in humans. Three isoforms of human endothelin have been identified: endothelin-1, -2, and -3. Endothelin-1 is a potent, 21-amino acid vasoconstrictor peptide produced by vascular endothelial cells. Endothelins are 21-amino acid vasoconstricting peptides produced primarily in the endothelium having a key role in vascular homeostasis. Endothelins are implicated in vascular diseases of several organ systems, including the heart, general circulation and brain. Endothelins are proteins that constrict blood vessels and raise blood pressure. They are normally kept in balance by other mechanisms, but when they are over-expressed, they contribute to high blood pressure(hypertension) and heart disease.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 16.5pg/ml, 7.8pg/ml, 3.9pg/ml human Endothelin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernatants, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human Endothelin standard solution and each sample is measured in duplicate.