Human MyD88 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0577
76235-694EA 760.6 USD
76235-694
Human MyD88 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human MyD88 AccuSignal ELISA Kit
  • Natural and recombinant human MyD88.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: M1-P296
Myeloid differentiation primary response gene(88)(MYD88) is a protein that, in humans, is encoded by the MYD88 gene. By interspecific backcross mapping, the mouse MyD88 gene was localized to chromosome 9; the human homolog was mapped to 3p22-p21.3 The human ortholog MYD88 seems to function similarly to mice, however, available evidence suggests that MYD88 is dispensable for human resistance to common viral infections and to all but a few pyogenic bacterial infections, demonstrating a major difference between mouse and human immune responses.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human MyD88 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates to each empty well. It is recommended that each human MyD88 standard solution and each sample be measured in duplicate.