Human THBS1 - TSP1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0540
76235-676EA 760.6 USD
76235-676
Human THBS1 - TSP1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human THBS1 - TSP1 AccuSignal ELISA Kit
  • Natural and recombinant human THBS1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: N19-P1170
Thrombospondin 1, also known as THBS1, is a protein that in humans in encoded by the THBS1 gene. Thrombospondin 1 is a subunit of a disulfide-linked homotrimeric protein. This protein is an adhesive glycoprotein that mediates cell-to-cell and cell-to-matrix interactions. By in situ hybridization, the THBS1 gene was mapped to human 15q15 and the cognate gene to mouse chromosome 2(region F) and was localized to 15q11-qter by Southern analysis of human-rodent somatic cell hybrids. Thrombospondin I is a multimodular secreted protein that associates with the extracellular matrix and possesses a variety of biologic functions, including a potent antiangiogenic activity. Other thrombospondin genes include thrombospondins II, III, and IV.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 100ng/ml, 50ng/ml, 25ng/ml, 1.25ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml human THBS1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human THBS1 standard solution and each sample is measured in duplicate.