Mouse CXCL10 - IP-10 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0430
76235-594EA 760.6 USD
76235-594
Mouse CXCL10 - IP-10 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CXCL10 - IP-10 AccuSignal ELISA Kit
  • Natural and recombinant mouse CXCL10.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: I22-P98
Chemokine(C-X-C motif) ligand 10(CXCL10) or IP-10 is a small cytokine belonging to the CXC chemokine family that is also known as 10 kDa interferon-gamma-induced protein(gamma-IP10 or IP-10). CXCL10 is secreted by several cell types in response to IFN-gamma. These cell types include monocytes, endothelial cells and fibroblasts. CXCL10 has been attributed to several roles, such as chemoattraction for monocytes/macrophages, T cells, NK cells, and dendritic cells, promotion of T cell adhesion to endothelial cells, antitumor activity, and inhibition of bone marrow colony formation and angiogenesis.It is a potent inhibitor of angiogenesis in vivo. The gene for CXCL10 is located on human chromosome 4 in a cluster among several other CXC chemokines. It is a RAS target gene and is overexpressed in the majority of colorectal cancers.This chemokine elicits its effects by binding to the cell surface chemokine receptor CXCR3.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse CXCL10 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma( heparin) to each empty well. It is recommended that each mouse CXCL10 standard solution and each sample be measured in duplicate.