Anti-AKT1 Mouse Monoclonal Antibody [Clone: 14E5.A2.B2.H9]

Supplier: Rockland Immunochemical
KCAI51
76235-518EA 463.88 USD
76235-518
Anti-AKT1 Mouse Monoclonal Antibody [Clone: 14E5.A2.B2.H9]
Antibodies
AKT1 Western Bloting kit allows for the detection of endogenous protein levels of human AKT1 present in cell lysates provided by the user. As a positive control, this kit includes a MDA-MB468 whole cell lysate proven to contain AKT1. After protein separation by SDS-PAGE and transfer, the membrane is probed with Rockland's optimized Anti-AKT1 antibody. Detection of the membrane bound antibody-antigen complex is achieved by the addition of a secondary antibody conjugated to the enzyme horseradish peroxidase. The enzyme reacts with a specialized formulation of luminol, an extremely sensitive, non-radioactive substrate that emits light and allows visualization using X-ray film or other imaging methods, including highly sensitive CCD cameras and imaging systems. AKT1 Western Bloting is ideal for investigators involved in Cell Signaling, Cancer, Neuroscience and Signal Transduction research.

Use Rockland Immunochemicals’ Anti-AKT1 Chemiluminescent Kit for Western Blotting for detection of human AKT1 native and recombinant proteins by western blot. Expect a band approximately 56 kDa in size corresponding to AKT1 protein. This kit is useful for both western blotting and dot blotting methods. Please read the entire product insert prior to use.

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 14E5.A2.B2.H9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR