HiTrap Columns, MabSelect PrismA, Cytiva
Supplier: Cytiva
17-5498-54 17-5498-51 17-5498-52 17-5498-53
76235-124EA 5007.61 USD
76235-124 76237-726 76237-728 76237-730
HiTrap Columns, MabSelect PrismA, Cytiva
Chromatography Columns
These columns are prepacked with MabSelect™ PrismA protein A chromatographyn resin that has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs. These columns are suitable for purifying antibodies with affinity chromatography in research and process development.
  • Enhanced binding capacity compared with other Protein A resins
  • Excellent alkaline stability enables efficient cleaning and sanitisation using 0.5 to 1.0 M NaOH
  • Convenient HiTrap™ format for easy connection to a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
MabSelect™ PrismA builds on the proven track record of MabSelect™ and MabSelect™ SuRe™ resins in commercial mAb production.
The columns are made of biocompatible PP that does not interact with biomolecules.
Caution: Note that HiTrap™ columns cannot be opened or repacked.