Anti-MAP2K1/MAP2K2 Mouse Monoclonal Antibody [Clone: 17C9.E1.H2.D7.H7.D5]

Supplier: Rockland Immunochemical
200-301-GZ6
76234-998EA 565.22 USD
76234-998
Anti-MAP2K1/MAP2K2 Mouse Monoclonal Antibody [Clone: 17C9.E1.H2.D7.H7.D5]
Antibodies
MEK1 pS222/MEK2 pS226 antibody detects MEK1 and MEK2. Mitogen-activated protein kinase kinase 1, (also known as MKK or MEK1), and Mitogen-activated protein kinase kinase 2, (also known as MEK2 or MKK2), are integral components of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. Activated MEK 1 and 2 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase. The MEK1 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.

Anti-MEK1 pS222/MEK2 pS226 (MOUSE) antibody is suitable for use in Western Blotting. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 43.5 kDa.

Type: Primary
Antigen: MAP2K1/MAP2K2
Clonality: Monoclonal
Clone: 17C9.E1.H2.D7.H7.D5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 lambda
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR