Anti-PARP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-GM8
76234-938EA 551.64 USD
76234-938
Anti-PARP1 Rabbit Polyclonal Antibody
Antibodies
PARP1 is the primary member of the poly(ADP-ribose) polymerase family, whose function is to signal DNA damage (and to recruit repair proteins) by PARylation. PARP1 is also involved in multiple cell death pathways, including apoptosis, necroptosis, autophagy, and a relatively new pathway termed parthanatos. It has been implicated in a new form of cell death termed parthanatos. PARP1 can also promote tissue survival by shifting the balance of cell death programs between autophagy and necrosis. Clinical studies have shown vulnerability to PARP inhibitors in DNA repair defective cancers. Anti-PARP1 (N-term ZF1) antibody is useful for researchers interested in cellular processes including DNA damage, transcriptional control, and stem cell identity research.

Anti-PARP1 (N-term ZF1) antibody has been validated by western blotting, IHC, and nanoimmunoassay (NIA). Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 113 kDa in size corresponding to PARP-1 by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: PARP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR