Anti-IKBKG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-GM7
76234-934EA 551.64 USD
76234-934
Anti-IKBKG Rabbit Polyclonal Antibody
Antibodies
Anti-NEMO antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Anti-NEMO antibody detects recombinant and endogenous NEMO. NEMO, the regulatory subunit of the IKK core complex, phosphorylates inhibitors of NF-kappa-B thus leading to the dissociation of the inhibitor/NF-kappa-B complex and ultimately the degradation of the inhibitor. Its binding to scaffolding polyubiquitin seems to play a role in IKK activation by multiple signaling receptor pathways. Nemo is also considered to be a mediator for TAX activation of NF-kappa-B and may be implicated in NF-kappa-B-mediated protection from cytokine toxicity. NEMO is essential for viral activation of IRF3 and involved in TLR3- and IFIH1-mediated antiviral innate response. The innate antivaral response from NEMO requires 'Lys-27'-linked polyubiquitination. Anti-NEMO is ideal for researchers interested in Immunology and Cancer research.

Anti-NEMO antibody is suitable for western blotting, immunoprecipitation, and ELISA. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 48kDa in size corresponding to endogenous NEMO protein by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: IKBKG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR