Anti-MAP2K2 Mouse Monoclonal Antibody (HRP) [Clone: 19G10.F1.E2]

Supplier: Rockland Immunochemical
200-303-GN8S
76234-918EA 150.22 USD
76234-918
Anti-MAP2K2 Mouse Monoclonal Antibody (HRP) [Clone: 19G10.F1.E2]
Antibodies
MEK2 antibodies detect the MEK2 isoform.  Mitogen-activated protein kinase kinase 2, also known as MEK2 or MKK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation.   This pathway also plays a key role in synaptic plasticity in the brain.  Activated MEK 2 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase.   MEK1 and MEK2 are about 80% identical to each other, and nearly identical within the kinase domain.   The MEK2 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.

Anti-MEK 2 HRP Conjugated (MOUSE) Antibody is suitable for use in Western Blotting and ELISA.  Specific conditions of reactivity should be optimized by the end user.  Expect a band of approximately 44 kDa.

Type: Primary
Antigen: MAP2K2
Clonality: Monoclonal
Clone: 19G10.F1.E2
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR