Anti-Slc2a2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-GN3
76234-784EA 551.64 USD
76234-784
Anti-Slc2a2 Rabbit Polyclonal Antibody
Antibodies
The Anti-Glut2 antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). The glucose transporter GLUT2 is a transmembrane carrier protein that allows protein facilitated glucose movement across cell membranes. GLUT2 is expressed in the plasma membranes of the liver, intestine, renal tubular cells, pancreatic islet beta cells, as well as in the portal and hypothalamic areas. Due to its low affinity and high capacity, GLUT2 transports dietary sugars, glucose, galactose and fructose in high concentrations, displaying large bidirectional fluxes in and out of cells. In pancreatic beta cells, GLUT2 is essential for glucose-stimulated insulin secretion. GLUT2 expression is necessary for the physiological control of glucose-sensitive genes, and its inactivation in the liver leads to impaired glucose-stimulated insulin secretion. In the nervous system, GLUT2-dependent glucose sensing regulates feeding, thermoregulation and pancreatic islet cell mass and function, as well as sympathetic and parasympathetic activities. In humans, inactivating mutations in GLUT2 cause Fanconi–Bickel syndrome, which is characterized by hepatomegaly and kidney disease. Anti-Glut2 is ideal for researchers interested in studying glucose transport mediated by Glut2 protein in the fields of diabetes, obesity, metabolism, and neuroscience research.

Glut-2 antibody has been tested for use in ELISA, immunohistochemistry, and by western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 57.1 kDa in size corresponding to Glut2 protein by western blotting in the appropriate stimulated tissue or cell lysate or extract.

Type: Primary
Antigen: Slc2a2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR