MOUSE SERUM (sterile) (Heat Inactivated), Rockland Immunochemicals
Supplier: Rockland Immunochemical
D108-10-0050 D108-10-0100 D108-10-0500 D108-10-1000
75933-960EA 1230.75 USD
75933-960 76226-474 76226-476 76226-478
MOUSE SERUM (sterile) (Heat Inactivated), Rockland Immunochemicals
Sera (Serum)
Heat Inactivated Sterile Serum
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Mouse Serum is used as a supplement to cell culture media. Mouse Serum is also suitable for use as a component of bioassays, immunoassays or enzyme assays. Mouse Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation. Mouse Serum is ideal for investigators in Cancer, Immunology, and Cell Biology research.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal