Brady® Workstation Scan and Print Suite CD, Brady Worldwide
Supplier: Brady Worldwide
BWS-SPS-CD
76226-066EA 2014.73 USD
76226-066
Brady® Workstation Scan and Print Suite CD, Brady Worldwide
Software Laboratory Software
Create production label workflows that you can pair with a barcode scanning device and printer.
  • Scan Mode - Scan existing barcodes to capture data and transfer onto a new label template
  • CSV Mode - Scan the name of a CSV file to print a specified batch of labels
  • Custom Script Mode - Perform advanced label operations such as parse scanned data or serialize labels
  • Scalable Workflows - Expand workflows to additional workstations on large projects using the Export and Import feature
  • Quickly replicate labels in a lab for sample testing, for aliquots at a bio bank, or in a variety of other environments
Save time and money by maximizing label printing output, reducing mistakes, and streamlining production workflows with the Brady Workstation Scan and Print Suite. This suite allows users to consume data from a barcode scanner, keyboard, or custom script to quickly populate custom label templates as part of a labeling workflow. Whether users need to replicate labels for lab specimens, re-label supplied parts for product assembly, or label products based off work orders, this software suite can solve any workflow needs. Be sure to pair the Scan and Print software with a barcode scanning device and Brady printer for the complete solution.
The Brady® Workstation Scan and Print Suite CD is compatible with BBP®11 Label Printer, BBP®12 Label Printer, BBP®31 Sign and Label Printer, BBP®33 Label Printer, BBP®35 Multi-Color Sign and Label Printer, BBP®37 Color & Cut Sign and Label Printer, BBP®81 Label Printer, BBP®85 Industrial Sign and Label Printer, BMP®41 Label Printer, BMP®51 Label Printer, BMP®53 Label Printer, BMP®61 Labeling System, BMP®71 Label Printer, BRADY IP® Series Label Printer, BradyJet™ J2000, BradyJet™ J5000, BradyPrinter i5100, BradyPrinter i7100, BradyPrinter S3100, BradyPrinter™ M611, GlobalMark® 2 Color and Cut Industrial Label Maker, GlobalMark® Multicolor Industrial Label Maker, IDXPERT™ Handheld Label Maker, J5000 Color Label Printer, LABXPERT™ Handheld Label Maker, PR300 Plus Industrial Label Printer, PR600 Plus Industrial Label Printer, TLS 2200® Thermal Labeling System, BradyPrinter i3300.