Human Recombinant MAP2K1
Supplier: Rockland Immunochemical
009-001-GQ6
76221-866EA 377.16 USD
76221-866
Human Recombinant MAP2K1
Proteins and Peptides
MEK1 double mutant human recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
MEK1 isoform double mutant recombinant protein is modified to put alanine residues at two key acitivation sites, Serine 218 and Serine 222. Mitogen-activated protein kinase kinase 1, also known as MKK or MEK, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. Activated MEK 1 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase. The inactive double mutant MEK1 recombinant protein is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.
MEK1 mutant protein is suitable as a negative control protein for immunoassays using antibodies targeting the critical MEK1 phosphorylated S218 or S222 residues. For western blot use at 50 ng or less. For other assays concentration is user optimized.