VWR® MatCapper Applicator
Supplier: VWR International
229078
76221-762EA 2908.87 USD
76221-762
VWR® MatCapper Applicator
Film Applicators Sealing Film Applicators
The MatCapper is a cap mat aplicator designed for use with storage and assay plates.
  • Caps shallow and deep well polypropylene storage plates
  • Requires minimal pressure for capping, reducing fatigue and RSI
  • Powder-coated to resist chemical spillage
  • Universal plate 'shuttle' accommodates shallow or deepwell plates
  • Equipped with fixing holes for securing to bench
For laboratories having to seal medium numbers of microplates, the VWR® MatCapper offers an affordable solution. Compact and portable, the MatCapper is very easy to use, requiring only one operation of the system to produce an accurate and tight seal on a wide range of both collection and shallow well microplates. The device is designed for use with EVA or sillicone mats, which work by friction fit and are re-usable.