VWR® Solid Bottom Assay Plates
Supplier: VWR International
76221-324
76221-324CS 610.25 USD
76221-324
VWR® Solid Bottom Assay Plates
Microplates HTS Microplates
These plates are designed to give optimum results from any instrument which reads from the top of the plate.
  • Black plates designed for top reading fluorescence instruments
  • Black plates have low background fluorescence and minimize light scattering
  • Highly polished wells give better readings
  • Alphanumerically labelled wells enable samples to be easily traced
Solid bottom 96-well assay plates is specifically designed for absorbance, fluorescence, luminescence, and scintillation applications. The design uses the most popular 96-well format with standard 'chimney' wells to overcome optical crosstalk and contamination. These plates are designed to give optimum results from any instrument which reads from the top of the plate.
Certifications: Complies with the standard ANSI/SLAS format.