Oakton® Automatic Temperature Compensation (ATC) Probes, Cole Parmer
Supplier: Antyila Scientific
35618-05
76221-270EA 98.37 USD
76221-270
Oakton® Automatic Temperature Compensation (ATC) Probes, Cole Parmer
Temperature Probes
Provides ATC for most current Oakton® pH meters.
  • Ensure optimal pH and ion measurement with ATC probes
Corrosion-resistant stainless steel thermistor features durable polymer handle and 3.3' (1 m) cable.