Oakton® Flexible Electrode Holder
Supplier: Antyila Scientific
35617-50
76221-268EA 408.71 USD
76221-268
Oakton® Flexible Electrode Holder
Electrode Holders
Keeps electrodes in position.
  • Weighted base pivots 360° for added convenience
Holds up to four standard size electrodes (14 mm dia. max.) and one ATC probe. Weighted base lets you pivot holder 360° for added convenience. Flexible arm lets you move electrode side to side or up and down while keeping electrode at a constant vertical angle.