Oakton Replacement Temperature Probe for 5+, 6+, 700, 2700 Meters, Cole Palmer
Supplier: Cole-Parmer
WD-35613-13
76221-246EA 76.55 USD
76221-246
Oakton Replacement Temperature Probe for 5+, 6+, 700, 2700 Meters, Cole Palmer
Temperature Probes
Provides ATC for most current Oakton pH meters.
Corrosion-resistant stainless steel 30 K thermistor features durable polymer handle and 3.3 ft (1-m) cable. Probes measure 88-mm L x 3-mm dia; handle adds another 67 mm to the total length. Temperature range is 0 to 100°C.