Oakton® EcoTestr™ CTS Pocket Meter, Cole-Parmer
Supplier: Antyila Scientific
35462-11
76221-244EA 104.5 USD
76221-244
Oakton® EcoTestr™ CTS Pocket Meter, Cole-Parmer
Electrochemistry Meters Conductivity Meters
The CTS meter is a reliable 3-in-1 tool that measures conductivity, TDS, and salinity.
  • Larger display with a better viewing angle
  • Automatic temperature compensation ensures accurate reading in varying environments
  • Easy-to-navigate interface for quick setup and calibration
  • Lanyard attachment makes field measurements more convenient
  • Battery level indicator flashes when battery is low
Expanded 4-digit dual-line display shows parameters and temperature readings simultaneously.
Redesigned leakproof cap functions as a sample cup and as a stabilizing base to stand the meter upright.
Rugged IP 67-rated housing is dustproof and waterproof and it floats.
Commonly used in agriculture, aquaculture and aquariums, cooling towers, education, environmental monitoring, food and beverage, hydroponics, manufacturing, pools and spas, printing, water treatment. Additionally EcoTestr CTS used in photo development and electroplating.
Delivery information: Four A76 1.5 V miniature alkaline batteries included.