Oakton® Ion 700 Benchtop pH and ISE Meter, Cole-Parmer
Supplier: OAKTON INSTRUMENTS MS
WD-35419-20 WD-35419-22 35419-23
101076-310EA 930 USD
101076-310 76221-218 76221-220
Oakton® Ion 700 Benchtop pH and ISE Meter, Cole-Parmer
Electrochemistry Meters pH and Multi-Parameter Meters
Designed for today's crowded laboratory benchtops.
  • Takes up less bench space—40% smaller than comparable models
  • Super-large dual-line display is easy to read
  • Simple calibrations with 5-point automatic pH buffer recognition for USA and NIST buffers
  • More accurate measurements with up to 5-point calibration for ion concentration on Ion 700 model
  • Versatile—use with any pH, ORP, or ISE electrodes with BNC connector
Ion 700 meter measures pH, mV, relative mV, and temperature, like a standard benchtop pH meter. When used with ISE's for ion concentration measurements, it also reads directly in ppm for easy use. Automatic temperature compensation (ATC) maintains reading accuracy even with fluctuating temperatures.
Meters feature Auto-Hold, selectable °F or °C measurement, and easy recall of electrode slope or offset.
Certifications: CE Approved; 3 Year Meter Warranty; ISO9001:2008.
Ordering information: Model WD-35419-20 includes meter with universal AC power adapter, one double-junction pH electrode, one temperature (ATC) probe, and a removeable electrode holder. For replacement parts, see model WD-35613-13 for the Automatic Temperature Compoensation (ATC) probe, model WD-35641-51 for a pH electrode, model WD-35615-07 for the universal power adapter, and model WD-35420-54 for the electrode holder.
Model WD-35419-22 includes meter with universal AC power adapter, and a removeable electrode stand. For replacement parts, see model WD-35615-07 for the univerrsal power adapter, and model WD-35420-54 for the electrode holder.
Model WD-35419-23 includes meter with universal AC power adapter, and one All-in-One pH/ATC electrode. For replacement parts, see model WD-35811-71 for the probe, model WD-35615-07 for the univerrsal power adapter, and model WD-35420-54 for the electrode holder.