Versaflo™ Powered Air Purifying Respirator (PAPR) Kit Easy Clean, 3M™
Supplier: 3M
7100150926
76220-762CS 3430.04 USD
76220-762
Versaflo™ Powered Air Purifying Respirator (PAPR) Kit Easy Clean, 3M™
Respirators Respiratory Systems
TR-800-ECK is a complete Powered Air Purifying Respirator featuring an intrinsically safe motor/blower. It includes everything required for use in operations where it is important to be able to easily clean the PAPR system.
  • Rated at Division I for Classes I, II, and III under the most current UL standard (UL 60079, 6th Edition, 2013)
  • Maintenance-free disposable hood saves cleaning time while providing a wide field of view
  • Choose from particulate filters or gas and vapor combination cartridges for protection in a variety of environments
  • Multiple airflow rates put users in control of personal comfort
  • Easy-to-clean belt saves time and effort, and storage and cleaning kit pave the way for more productivity
The TR-800 is a motor/blower certified by UL with an intrinsically safe (IS) rating of Division 1: IS Class 1, II, III; Division 1 (includes Division 2) Groups C, D, E, F, G; T4, under the most current standard (UL 60079, 6th Edition, 2013).
Delivery information: The kit includes S-433L Hood with Integrated Head Suspension, BT-30 Length Adjusting Breathing Tube, TR-6710N (HE) Filter, TR-641N Single Station Battery Charger, BT-922 Breathing Tube Cover, Intrinsically Safe TR-802N PAPR Unit, TR-830 Intrinsically Safe Battery, TR-838 Battery Attachment Tool, TR-627 Easy Clean Belt, TR-6700 Filter Cover, TR-6600 Prefilter (X2), TR-662 Spark Arrestor (X2), BT-922 Breathing Tube Cover, TR-653 Storage and Cleaning Kit, and TR-971 Airflow Indicator.