2,6-Difluoro-3-iodopyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

D5141-1G D5141-5G
76219-176EA 57 USD
76219-176 76219-178
2,6-Difluoro-3-iodopyridine ≥98.0% (by GC)
2,6-Difluoro-3-iodopyridine
Formula: C₅H₂F₂IN
MDL Number: MFCD09743595
CAS Number: 685517-67-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR