6-Chloropyridine-2,3-diamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D4765-1G
76219-070EA 82 USD
76219-070
6-Chloropyridine-2,3-diamine ≥98.0% (by GC, titration analysis)
6-Chloropyridine-2,3-diamine
Formula: C₅H₆ClN₃
MDL Number: MFCD00209966
CAS Number: 40851-95-4

Order Now

Specification Test Results

Appearance White to Brown to Dark purple powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 132.0 to 136.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR