(R)-(+)-2,2'-Diamino-1,1'-binaphthalene ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

D3207-200MG
76219-018EA 42 USD
76219-018
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene ≥98.0% (by HPLC, titration analysis)
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene
Formula: C₂₀H₁₆N₂
MDL Number: MFCD00145204
CAS Number: 18741-85-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 240.0 to 244.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR