Trubore Pressure Bearing with Ace-Threds, 19 mm, Ace Glass
Supplier: Ace Glass
8049-02
76214-466EA 931.74 USD
76214-466
Trubore Pressure Bearing with Ace-Threds, 19 mm, Ace Glass
Stirrers Stirring Shafts
Glass bearing designed for use with Ace-Thred pressure reactors and ACE polished glass 19 mm stir shafts.
  • Bearing made of high-quality borosilicate glass
  • Both the top bushing and bottom coupling are PTFE
  • Maximum pressure is 30 psig at ambient temperature
  • Maximum rpm: 350