LopHene® Ⅱ Concentrated Low-pH Disinfectant, Decon Labs
Supplier: Decon Labs
8811
76214-018CS 478.16 USD
76214-018
LopHene® Ⅱ Concentrated Low-pH Disinfectant, Decon Labs
Cleaning Agents Disinfectants
LopHene® Ⅱ concentrated phenolic germicide for rotational disinfection.
  • High soil-load and hard water tolerance
  • Diluted one ounce per gallon of water (1:128)
It can be used on hard surfaces in labs, production areas and on process equipment or anywhere thorough disinfection efficacy is required. LopHene II is a biological decontamination product and disinfection. It is effective against a wide range of bacteria, viruses and fungi, including TB and HIV-1.
Certifications: Conforms to OSHA’s blood borne Pathogens Standard. EPA registered.
Ordering information:
Available in convenient packaging configurations. Custom solutions or packaging configuration may be available. Please note that pure ethanol is regulated by the US ATF/TTB and depending on the taxable status of your institution, pure ethanol products are taxed according to the government schedule of taxes detailed in 27 CFR Chapter 1 section 27.41. To purchase tax-free, you must have an Industrial Alcohol User Permit from the AFT/TTB, specifically marked for “the withdraw and use of alcohol free of tax”. Tax-free pure ethanol is intended for industrial use only.