Dissolved Oxygen Meter Kit, Sper Scientific
Supplier: Sper Scientific
850081DOK
76214-014EA 937.82 USD
76214-014
Dissolved Oxygen Meter Kit, Sper Scientific
Electrochemistry Meters Oxygen Meters
Measures dissolved oxygen to determine water quality in environmental testing, labs, industrial and municipal waste water, aquariums and fish hatcheries.
  • Reliable DO readings at a reasonable price
  • Features simple one-button calibration to air
The back-lit display simultaneously displays dissolved oxygen (in % or mg/l) and temperature readings (in °C or °F). Automatic temperature compensation as well as manual compensation for salinity and altitude. DO probe heads include built in membranes and screw on and off for easy replacement. Also functions as a pH, ORP, conductivity, TDS or salinity meter when used with the optional probes.
Delivery information: Comes in a hard carrying case complete with DO probe, two spare probe heads, electrolyte, instructions and a 9 V battery. EPA equivalency method: 360.1.