Acid Fast Stain Kit, Kinyoun, Cold Method for AFB, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
AF900
76213-940EA 58.41 USD
76213-940
Acid Fast Stain Kit, Kinyoun, Cold Method for AFB, Hardy Diagnostics
Stains Microbiology Stains
The kit is used as a differential method to detect the difference between those organisms which decolorize with treatment of acid alcohol, and those that do not, such as the mycobacteria.
This is the “cold stain” method that does not require heating of the slide.
Delivery information: Kit includes one 8 oz. bottle of each, carbol fuchsin (kinyoun), acid alcohol (decolorizer) and methylene blue (counterstain).