Ethanol 70%, meets analytical specification of USP (140 Proof)

Supplier: Decon Labs
Danger

2432
76212-358CS 181.44 USD
76212-358
Ethanol 70%, meets analytical specification of USP (140 Proof)
Ethanol

This 140 Proof ethanol (70% ethanol) is a pure ethyl alcohol made for research, healthcare, industrial, or production applications as a cleaner, solvent, or reagent.


Composition: Ethyl alcohol: 70%, Purified water: 30%


Certifications: Meets USP specifications. "Industrial Alcohol User Permit" issued by TTB.

Formula: C₂H₅OH
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,III

Order Now

Specification Test Results

Assay-Ethyl Alcohol (%) 68.0 - 72.0
Specific Gravity @ 15.56 0.8848 - 0.8948
Water (%) 28.0 - 32.0
Color Passes USP Test for Color
Clarity Passes USP Test for Clarity
Acidity 30
Alkalinity Passes USP Test
Non-volatile Residue =2.5
Volatile Impurities (Acetaldehyde) =10
Volatile Impurities (Benzene) =2
Volatile Impurities (Methanol) =200
Volatile Impurities (Other Impurities) =300
UV Absorbance Curve at 235 - 340 nm run against Water Passes USP for Smooth Curve
UV Absorbance at 240 nm =0.40
UV Absorbance at 250 nm =0.30
UV Absorbance at 260 nm =0.30
UV Absorbance at 270 - 340 nm =0.10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR