Zymoprep™ Yeast Plasmid Miniprep II, Zymo Research
Supplier: Zymo Research
D2004
76212-310KT 209.07 USD
76212-310
Zymoprep™ Yeast Plasmid Miniprep II, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Zymoprep™ Yeast Plasmid Miniprep II used for plasmid isolation from S. pombe.
  • Simple and Efficient
  • No need for glass beads or phenol
System is ideal for low-copy number and hard to isolate plasmids. Eluted plasmid DNA can be used directly for E. Coli transformation, PCR, and Southern blot analysis.
cerevisiae, C. albicans and S. pombe, and any fungi whose cell walls are susceptible to yeast lytic enzyme lysis.