Scoop/Ladle, HDPE, Dynalon
Supplier: Dynalon
108085-50
76210-886CS 270.23 USD
76210-886
Scoop/Ladle, HDPE, Dynalon
Scoops
Plastic ladle dippers with a heavy-duty bowl for sampling and dispensing corrosive liquids.
  • Handy for sampling and dispensing corrosive liquids
Fabricated in the USA from high density polyethylene, these plastic ladle dippers feature a high-temperature, extra heavy-duty bowl with securely welded solid handle.