VWR® OTG Safety Glasses
Supplier: VWR International
OTG-R
76210-732CS 1637.73 USD
76210-732
VWR® OTG Safety Glasses
Safety Glasses
Economical and compact, designed to fit over prescription glasses.
  • Fits over most prescription glasses
  • Adjustable temple length for varied facial contours
  • Polycarbonate lens
  • Provides 99.9% UV protection
The pantoscopic adjustment allows lens angle modification and it is impact-resistant.
Certifications: Meets ANSI Z87.1 standards.