Jar with Cap, HDPE, Dynalon
Supplier: Dynalon
226415-1500
76210-710PK 149.08 USD
76210-710
Jar with Cap, HDPE, Dynalon
Jars General Use Jars
These cylindrical food-grade HDPE containers with black HDPE screw closures are supplied with a neck insert for added security.
They are suitable for powders, solid specimens, creamy or semisolid substances, crystals, and hygroscopic substances.
Certifications: Suitable for foodstuff according to EC Regulation 10/2011.