Savant™ SpeedVac™ Modular Vacuum Concentrator, Thermo Scientific
Supplier: Thermo Fisher Scientific
SPD130P1-115
76210-592EA 29300 USD
76210-592
Savant™ SpeedVac™ Modular Vacuum Concentrator, Thermo Scientific
Evaporators Centrifugal Evaporators
The Thermo Scientific™ Savant™ SpeedVac™ SPD130DLX vacuum concentrator is a medium-capacity, vacuum concentrator with a modular design.
  • Kit
  • Medium capacity
  • Variety of rotors
  • Radiant glass cover
  • PTFE coated aluminum chamber
The SpeedVac™ SPD130DLX is compatible with a wide variety of solvents and offers resistance to TFA, DMSO, and other aggressive solvents used in combinatorial chemistry applications.
Ordering information: Supplied as a package, please see description for details of what is included in kit/package.